Learn More
Nickel phthalocyanine, 95%
CAS: 14055-02-8 | C32H16N8Ni | 571.23 g/mol
$128.13 - $128.13
Chemical Identifiers
| CAS | 14055-02-8 |
|---|---|
| Molecular Formula | C32H16N8Ni |
| Molecular Weight (g/mol) | 571.23 |
| MDL Number | MFCD00041954 |
| InChI Key | LVIYYTJTOKJJOC-UHFFFAOYSA-N |
| Synonym | nickel phthalocyanine, nickel ii phthalocyanine, phthalocyanine, ni, nickel ii phthalocyanine, dye content 85 % |
| PubChem CID | 2735100 |
| IUPAC Name | nickel(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹â°.1¹²,¹â¹.1²¹,²â¸.0â´,â¹.0¹³,¹â¸.0²²,²â·.0³¹,³â¶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide |
| SMILES | [Ni++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC415600050
|
Thermo Scientific Chemicals
415600050 |
5 g | Glass bottle |
Each for $128.13
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14055-02-8 | |
| 571.23 | |
| LVIYYTJTOKJJOC-UHFFFAOYSA-N | |
| 2735100 | |
| [Ni++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31 |
| C32H16N8Ni | |
| MFCD00041954 | |
| nickel phthalocyanine, nickel ii phthalocyanine, phthalocyanine, ni, nickel ii phthalocyanine, dye content 85 % | |
| nickel(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹â°.1¹²,¹â¹.1²¹,²â¸.0â´,â¹.0¹³,¹â¸.0²²,²â·.0³¹,³â¶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide |
Specifications
| 14055-02-8 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| MFCD00041954 | |
| nickel phthalocyanine, nickel ii phthalocyanine, phthalocyanine, ni, nickel ii phthalocyanine, dye content 85 % | |
| [Ni++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31 | |
| 571.23 | |
| 571.21 | |
| Crystalline Powder |
| 94.0 | |
| Blue | |
| 95% | |
| C32H16N8Ni | |
| 5 g | |
| LVIYYTJTOKJJOC-UHFFFAOYSA-N | |
| nickel(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹â°.1¹²,¹â¹.1²¹,²â¸.0â´,â¹.0¹³,¹â¸.0²²,²â·.0³¹,³â¶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide | |
| 2735100 | |
| 95% | |
| Nickel phthalocyanine, 95% |
Safety and Handling
GHS H Statement:
Suspected of causing cancer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 237-893-3
TSCA : TSCA
RUO – Research Use Only