missing translation for 'onlineSavingsMsg'
Learn More

Nickel phthalocyanine, 95%

Catalog No. AC415600050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415600050 5 g Glass bottle
AC415600250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC415600050 Supplier Thermo Scientific Chemicals Supplier No. 415600050

CAS: 14055-02-8 | C32H16N8Ni | 571.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14055-02-8
Molecular Formula C32H16N8Ni
Molecular Weight (g/mol) 571.23
MDL Number MFCD00041954
InChI Key LVIYYTJTOKJJOC-UHFFFAOYSA-N
Synonym nickel phthalocyanine, nickel ii phthalocyanine, phthalocyanine, ni, nickel ii phthalocyanine, dye content 85 %
PubChem CID 2735100
IUPAC Name nickel(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
SMILES [Ni++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Blue
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Formula Weight 571.21
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Nickel phthalocyanine, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Suspected of causing cancer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 237-893-3
TSCA TSCA

RUO – Research Use Only