missing translation for 'onlineSavingsMsg'
Learn More

Nickel Nitrate Hexahydrate, ACS, 98-102%, Spectrum™ Chemical

Catalog No. p-7165961
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Catalog No. Quantity Packaging
18611972 25 g Amber Glass Bottle
18611973 125 g Amber Glass Bottle
18611974 500 g Amber Glass Bottle
18611975 2.5 kg Amber Glass Bottle
18611976 45 kg Fiber Drum
5 options

Catalog No. 18611972

Supplier: Spectrum Chemical Mfg Cor N126725GM

Only null left

Chemical Identifiers

CAS 13478-00-7
Molecular Formula H12N2NiO12
Molecular Weight (g/mol) 290.79
MDL Number MFCD00149805
InChI Key AOPCKOPZYFFEDA-UHFFFAOYSA-N
IUPAC Name nickel(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98 to 102%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.