missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Nitrate, Crystal, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor N106212KG
Specifications
| 13478-00-7 | |
| 12 kg | |
| MFCD00149805 | |
| O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 290.79 | |
| Poly Pail |
| 1 | |
| H12N2NiO12 | |
| AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
| nickel(2+) hexahydrate dinitrate | |
| Reagent |
Chemical Identifiers
| 13478-00-7 | |
| 290.79 | |
| AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
| O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| H12N2NiO12 | |
| MFCD00149805 | |
| nickel(2+) hexahydrate dinitrate |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.