missing translation for 'onlineSavingsMsg'
Learn More

Nickel Iron, sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis), Thermo Scientific™

Catalog No. AA40887KS
Change view
Click to view available options
Quantity:
1 ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40887KS 1 ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40887KS Supplier Thermo Scientific Chemicals Supplier No. 40887KS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Name or Material Nickel Iron, sputtering target
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Avg. Mol. Wt. or Mol. Wt. Range Ni:Fe; 81:19 wt%
Quantity 1 ea.
Assay Percent Notes (metals basis)
MDL Number MFCD01741928
Percent Purity 99.95%