missing translation for 'onlineSavingsMsg'
Learn More

Nickel (II) Sulfate Hexahydrate, Purified, Reagents

Catalog No. p-200021240
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337157 100 g
01337156 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337157 Supplier Labchem Inc Supplier No. C2256500100C2

Nickel (II) Sulfate Hexahydrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.84
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym Nickelous Sulfate Hexahydrate, Nickel (II) Sulfate Hexahydrate
IUPAC Name nickel(2+) hexahydrate sulfate
SMILES O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

Specifications

Boiling Point 103°C
Color Blue to Green
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 3.68
Chemical Name or Material Nickel (II) Sulfate Hexahydrate, Purified