missing translation for 'onlineSavingsMsg'
Learn More

Nickel (II) Sulfate Hexahydrate, Purified, Reagents

Catalog No. 01337157
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337157 100 g
01337156 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337157 Supplier Labchem Inc Supplier No. C2256500100C2
Only null left

Nickel (II) Sulfate Hexahydrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.84
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym Nickelous Sulfate Hexahydrate, Nickel (II) Sulfate Hexahydrate
IUPAC Name nickel(2+) hexahydrate sulfate
SMILES O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

Specifications

Boiling Point 103°C
Color Blue to Green
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 3.68
Chemical Name or Material Nickel (II) Sulfate Hexahydrate, Purified