missing translation for 'onlineSavingsMsg'
Learn More

Nickel(Ii) Sulfate Hexahydrate, ACS Reagent, Solstice

Catalog No. 6001083
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001083 1 kg
6001084 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001083 Supplier Solstice Supplier No. 2276761KG

ACS Reagent, 99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.839
MDL Number MFCD00149813
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
PubChem CID 5284429
ChEBI CHEBI:53437
IUPAC Name nickel(2+);sulfate;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

Specifications

Quantity 1 kg
Linear Formula NiSO4 · 6H2O
UN Number 3077
Formula Weight 262.85g/mol
Percent Purity 99%
Grade ACS Reagent
Density Vapor Density: 2.07 (vs air)
Chemical Name or Material Nickel(Ii) Sulfate Hexahydrate
Hazard Statement P201-P261-P273-P280-P284-P304 + P340 + P312
Precautionary Statement H302 + H332-H315-H317-H334-H341-H350i-H360D-H372-H410
EINECSNumber 232-104-9
RTECSNumber QR9600000