missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) sulfate hexahydrate, ACS, 98.0% min

Catalog No. p-4407675
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3633622 100 g
AA3633636 500 g
AA36336A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3633622 Supplier Thermo Scientific Chemicals Supplier No. 03633622
Only null left

CAS: 10101-97-0 | H12NiO10S | 262.839 g/mol

It is the precursor material for Ni electroplating. It is applied in biochemistry and also employed as a calibrant for magnetic susceptibility measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.839
MDL Number MFCD00149813
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
PubChem CID 5284429
ChEBI CHEBI:53437
IUPAC Name nickel(2+);sulfate;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

Specifications

Color Blue
Physical Form Powder
Quantity 100 g
Assay Percent Range 98% min.
Solubility Information Soluble in water. Slightly soluble in alcohol and methanol
Formula Weight 262.86 (154.77 Anhydrous)
Grade ACS
Density 2.07
Chemical Name or Material Nickel (II) Sulfate Hexahydrate, For analysis ACS
Hazard Category H302-H315-H317-H319-H332-H334-H335-H341-H350i-H360D-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
DOTInformation DOT Class: 9; Packaging Group: III
EINECSNumber 232-104-9
RTECSNumber QR9600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only