missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) sulfate hexahydrate, ≥99.99% trace metals basis, MilliporeSigma™ Supelco™

Catalog No. 111018989
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018989 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018989 Supplier Merck Emd Millipore Supplier No. 46790150G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity grade (≥99.99% trace metals basis) suitable for laboratory research and analytical applications.

Chemical Identifiers

Molecular Weight (g/mol) 262.85
MDL Number MFCD00149813

Specifications

Physical Form Solid
CAS 10101-97-0
Quantity 50 g
Linear Formula NiSO4 · 6H2 O
Formula Weight 262.85
Percent Purity ≥99.99% (trace metals basis)
Packaging Poly Bottle
Density Vapor Density: 2.07 (vs air)
Chemical Name or Material Nickel(II) Sulfate Hexahydrate
RTECSNumber QR9600000