missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) sulfate hexahydrate, 98%

Catalog No. p-7049019
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1251436 500 g
AA12514A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1251436 Supplier Thermo Scientific Chemicals Supplier No. 01251436

In nickel plating, as mordant in dyeing and in printing textiles | CAS: 10101-97-0 | H12NiO10S | 262.839 g/mol

Nickel sulfate hexahydrate is mainly used in Ni plating, as mordant in dyeing and in printing textiles, as a blackening agent for zinc and brass and in the fabrication of organic nickel salts. It is used as an effective catalyst for gas-phase dehydration of glycerol to acrolein. Nickel sulfate hexahydrate crystal is an efficient ultraviolet filter material. Columns used in biochemistry and molecular biology can be regenerated with nickel sulfate. It is used to prepare nickel carbonate, a precursor to pigments and nickel-based catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.839
MDL Number MFCD00149813
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
PubChem CID 5284429
ChEBI CHEBI:53437
IUPAC Name nickel(2+);sulfate;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

Specifications

Physical Form Crystalline
Quantity 500 g
Assay Percent Range 98%
Linear Formula NiSO4·6H2O
UN Number UN3288
Merck Index 14,6517
Solubility Information Very soluble in water,ethanol,ammonia.
Formula Weight 262.86 (154.77 Anhydrous)
Percent Purity 98%
Density 2.07 g/mL
Chemical Name or Material Nickel(II) sulfate hexahydrate
Show More Show Less
Hazard Category H302-H315-H317-H319-H332-H334-H335-H341-H350i-H360D-H372
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H315-H317
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 232-104-9
RTECSNumber QR9600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only