Learn More
Nickel(II) sulfate hexahydrate, 98+%, ACS reagent
CAS: 10101-97-0 | NiO4S·6H2O | 262.85 g/mol
Supplier: Thermo Scientific Chemicals 415610051
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Nickel(II) sulfate hexahydrate | |
| 10101-97-0,7786-81-4 | |
| 98.0 | |
| Crystals | |
| 98.0 to 102% (Complexometry) | |
| NiSO4·6H2O | |
| 3288 | |
| nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
| RRIWRJBSCGCBID-UHFFFAOYSA-L | |
| nickel(2+);sulfate;hexahydrate | |
| 5284429 | |
| 262.85 | |
| ACS Reagent |
| 53.0°C | |
| Blue-Green | |
| 100.0 | |
| 5 kg | |
| NiO4S·6H2O | |
| MFCD00149813 | |
| 15,6602 | |
| Solubility in water: 625g/L (20°C). Other solubilities: sparingly soluble in alcohol | |
| O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] | |
| 262.85 | |
| CHEBI:53437 | |
| ≥98% | |
| Plastic Bottle |
Chemical Identifiers
| 10101-97-0 | |
| 262.85 | |
| RRIWRJBSCGCBID-UHFFFAOYSA-L | |
| 5284429 | |
| nickel(2+);sulfate;hexahydrate |
| NiO4S·6H2O | |
| MFCD00149813 | |
| nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
| CHEBI:53437 | |
| O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Suspected of causing genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
M
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
GHS Signal Word: Danger
RTECSNumber : QR9600000
RUO – Research Use Only