Learn More
Nickel(II) sulfate, 0.5M aq. soln., Thermo Scientific Chemicals
$151.45 - $151.45
Chemical Identifiers
| CAS | 10101-97-0 |
|---|---|
| Molecular Formula | H12NiO10S |
| Molecular Weight (g/mol) | 262.839 |
| MDL Number | MFCD00011146 |
| InChI Key | RRIWRJBSCGCBID-UHFFFAOYSA-L |
| Synonym | nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 |
| PubChem CID | 5284429 |
| ChEBI | CHEBI:53437 |
| IUPAC Name | nickel(2+);sulfate;hexahydrate |
| SMILES | O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ61993AE
|
Thermo Scientific Chemicals
J61993AE |
100 mL |
Each for $151.45
|
|
|||||
Description
Nickel(II) sulfate, 0.5M aq. soln. is used in nickel plating, as mordant in dyeing and in printing textiles. Columns used in polyhistidine-tagging, useful in biochemistry and molecular biology, are regenerated with nickel sulfate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNickel(II) sulfate, 0.5M aq. soln. is used in nickel plating, as mordant in dyeing and in printing textiles. Columns used in polyhistidine-tagging, useful in biochemistry and molecular biology, are regenerated with nickel sulfate.
Solubility
Fully miscible in water.
Notes
Protect from heat.
Chemical Identifiers
| 10101-97-0 | |
| 262.839 | |
| RRIWRJBSCGCBID-UHFFFAOYSA-L | |
| 5284429 | |
| nickel(2+);sulfate;hexahydrate |
| H12NiO10S | |
| MFCD00011146 | |
| nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
| CHEBI:53437 | |
| O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Specifications
| 10101-97-0 | |
| Liquid | |
| H12NiO10S | |
| MFCD00011146 | |
| nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
| RRIWRJBSCGCBID-UHFFFAOYSA-L | |
| nickel(2+);sulfate;hexahydrate | |
| 5284429 | |
| 262.86 | |
| Odorless |
| Green | |
| 100 mL | |
| NiSO4 | |
| UN3082 | |
| Fully miscible in water. | |
| O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] | |
| 262.839 | |
| CHEBI:53437 | |
| 0.5M aq. soln. | |
| Nickel(II) sulfate |
Safety and Handling
GHS H Statement
H334-H350-H360-H372-H341-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes skin irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280-P281-P285-P302+P352-P304+P341-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H341-H350i-H360D-H372
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only