missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) sulfamate, 50% w/w aq. soln., Reagent Grade
Description
Nickel plating Nickel(II) sulfamate is used for electroplating and as a surface treating agent. It finds application in metal coloring and casting.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Green |
| Physical Form | Liquid |
| Quantity | 1 L |
| Odor | Odorless |
| Linear Formula | Ni(SO3NH2)2 |
| UN Number | UN3082 |
| Solubility Information | Miscible with water,immiscible with ether,acetone. |
| Formula Weight | 250.86 (Anhydrous) |
| Grade | Reagent |
| Concentration or Composition (by Analyte or Components) | 50% w/w aq. soln. |
| Show More |
Safety and Handling
| Hazard Category | H317-H334-H341-H350i-H360D-H372 |
| Hazard Statement | GHS H Statement H334-H350-H360-H372-H341-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S. |
| EINECSNumber | 237-396-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only