missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) phthalocyanine

Catalog No. p-7050784
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3945306 5 g
AA3945314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3945306 Supplier Thermo Scientific Chemicals Supplier No. 03945306

CAS: 14055-02-8 | C32H16N8Ni | 571.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14055-02-8
Molecular Formula C32H16N8Ni
Molecular Weight (g/mol) 571.23
MDL Number MFCD00041954
InChI Key LVIYYTJTOKJJOC-UHFFFAOYSA-N
Synonym nickel phthalocyanine, nickel ii phthalocyanine, phthalocyanine, ni, nickel ii phthalocyanine, dye content 85 %
PubChem CID 2735100
IUPAC Name nickel(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
SMILES [Ni++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31

Specifications

Melting Point >300°C
Color Blue to Purple
Quantity 5 g
Formula Weight 571.24
Physical Form Powder
Chemical Name or Material Nickel(II) phthalocyanine
Hazard Category H317-H334-H351
Hazard Statement GHS H Statement
H350-H302-H312-H332
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P261-P272-P280-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
EINECSNumber 237-893-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only