missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) perchlorate hexahydrate, Reagent Grade

Catalog No. AA1163114
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1163114 25 g
AA1163122 100 g
AA1163136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1163114 Supplier Thermo Scientific Chemicals Supplier No. 01163114

CAS: 13520-61-1 | Cl2H12NiO14 | 365.675 g/mol

Nickel(II) perchlorate hexahydrate is used as a starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13520-61-1
Molecular Formula Cl2H12NiO14
Molecular Weight (g/mol) 365.675
MDL Number MFCD00149804
InChI Key WHGYCGOFTBFDLW-UHFFFAOYSA-L
Synonym nickel ii perchlorate hexahydrate, acmc-1boln, 2clo4.ni.6h2o, nickel ii perchlorate hydrate, nickel 2+ hexahydrate diperchlorate, nickel perchlorate, hexahydrate, reagent, nickel ii perchlorate hexahydrate, reagent grade, nickel ii perchlorate hexahydrate trace metals basis, nickel ii perchlorate hexahydrate, purum p.a., crystallized kt
PubChem CID 15769463
IUPAC Name nickel(2+);diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ni+2]

Specifications

Melting Point 140°C
Color Green
Physical Form Damp Crystalline Solid
Quantity 25 g
Solubility Information Very soluble in water. Soluble in many organic solvents
Formula Weight 365.70 (257.59 Anhydrous)
Grade Reagent
Odor Odorless
Refractive Index 1.55
Sensitivity Hygroscopic
Appearance Green crystals
Chemical Name or Material Nickel(II) perchlorate hexahydrate
Show More Show Less
Hazard Category H272-H314-H317-H334-H335-H341-H350-H360-H372
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 237-124-1
RTECSNumber SC9550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only