missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) octanoate, in mineral spirits (8% Ni)

Catalog No. AA3947914
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3947914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3947914 Supplier Thermo Scientific Chemicals Supplier No. 03947914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4995-91-9 | C8H16NiO2 | 202.907 g/mol

Nickel(II) octanoate is used to prepare nickel based catalysts using niobia modified silica as support. It is also used as a catalyst and involved in the treatment of qaiyarah heavy crude oil, petrolene using ethanol as a solvent and reductant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4995-91-9
Molecular Formula C8H16NiO2
Molecular Weight (g/mol) 202.907
MDL Number MFCD00064239
InChI Key ANYMYNDNCKURBB-UHFFFAOYSA-N
Synonym nickel dioctanoate, nickel 2+ caprylate, nickel octoate, caprylic acid, nickel ii salt, octanoic acid, nickel 2+ salt
PubChem CID 54611835
IUPAC Name nickel;octanoic acid
SMILES CCCCCCCC(=O)O.[Ni]

Specifications

Color Green
Density 0.95 g/mL
Flash Point 40°C (104°F)
Linear Formula [CH3(CH2)6COO]2Ni
Quantity 25 g
UN Number UN1993
Solubility Information Miscible with ethanol,chloroform,acetonitrile,ether,carbon disulfide,petroleum ether and glacial acetic acid. Immiscible with water.
Formula Weight 345.1
Concentration or Composition (by Analyte or Components) 8% Ni
Physical Form Liquid
Chemical Name or Material Nickel(II) octanoate, In mineral spirits
Show More Show Less
Hazard Category H226-H304-H317-H334-H340-H350i-H360D-H372
Hazard Statement GHS H Statement
H334-H340-H350-H360-H372-H304-H226-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P272-P280g-P281-P285-P301+P310-P303+P361+P353-P304+P341-P308+P313-P331-P333+P313-P342+P311-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only