missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Nitrate Hexahydrate, purum p.a, ≥97.0% (KT), Honeywell™

Catalog No. p-7120621
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
Catalog No. Quantity Packaging
60047426 250 g Poly bottle
60047425 1 kg Poly bottle
2 options

Catalog No. 60047426

Supplier: Honeywell Chemicals 72253250G

Only null left

Purum p.a., crystallized,≥97.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Chemical Identifiers

CAS 13478-00-7
Molecular Formula H12N2NiO12
Molecular Weight (g/mol) 290.79
MDL Number MFCD00149805
InChI Key AOPCKOPZYFFEDA-UHFFFAOYSA-N
Synonym nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
PubChem CID 61630
SMILES O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 56.7°C
Color Blue Green
pH 4.0 (at 20°C)
Physical Form Solid
Quantity 250 g
Linear Formula Ni(NO3)2 · 6H2O
UN Number UN2725
Solubility Information 2.385g/L at 20°C
IUPAC Name nickel(2+) hexahydrate dinitrate
Formula Weight 290.79g/mol
Percent Purity ≥97.0% (KT)
Packaging Poly bottle
Density 2.05g/mL at 25°C
Chemical Name or Material Nickel(II) nitrate hexahydrate
Show More Show Less
Hazard Statement P201-P210-P261-P273-P280-P284
DOTInformation 5.1 / PGIII
RTECSNumber 236-068-5