missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) hydroxide, for analysis

Catalog No. p-43704
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC275022500 250 g Glass bottle
AC275020010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC275022500 Supplier Thermo Scientific Chemicals Supplier No. 275022500
Only null left

CAS: 12054-48-7 | H2NiO2 | 92.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 12054-48-7
Molecular Formula H2NiO2
Molecular Weight (g/mol) 92.71
MDL Number MFCD00011140
InChI Key BFDHFSHZJLFAMC-UHFFFAOYSA-L
IUPAC Name nickel(2+) dihydroxide
SMILES [OH-].[OH-].[Ni++]

Specifications

Melting Point 230.0°C
Color Green
Physical Form Crystalline Powder
Quantity 250 g
Linear Formula Ni(OH)2
Merck Index 15, 6595
Solubility Information Solubility in water: 0.13g/L (20°C). Other solubilities: soluble in dil. acids,ammonia
Formula Weight 92.7
Percent Purity 61.0 to 65.0% (Nickel)
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Nickel(II) hydroxide, c.p., 60-61% Ni
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage the unborn child.
Suspected of causing genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENT
EINECSNumber 235-008-5
RTECSNumber QR7040000
TSCA TSCA

RUO – Research Use Only