Learn More
Nickel(II) formate dihydrate
For Nickel Catalyst Preparations | CAS: 15694-70-9 | Ni(HCO2)2·2H2O
$87.90 - $1242.78
Chemical Identifiers
| CAS | 15694-70-9 |
|---|---|
| Molecular Formula | Ni(HCO2)2·2H2O |
| MDL Number | MFCD00153034 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1380122
|
Thermo Scientific Chemicals
01380122 |
100 g |
Each for $87.90
|
|
|||||
|
AA1380136
|
Thermo Scientific Chemicals
01380136 |
500 g |
Each for $349.53
|
|
|||||
|
AA13801A3
|
Thermo Scientific Chemicals
013801A3 |
2 kg |
Each for $1,242.78
|
|
|||||
Description
Nickel(II) formate dihydrate is used in nickel catalyst preparations. It is also used as a biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15694-70-9 | |
| Ni(HCO2)2·2H2O | |
| MFCD00153034 |
Specifications
| 130°C to 140°C-2H2O | |
| 15694-70-9 | |
| 100 g | |
| MFCD00153034 | |
| 184.77 (148.74 Anhydrous) | |
| In the range 180° to 200°C decomposes to Ni,CO,CO2 | |
| Nickel(II) formate dihydrate |
| Green | |
| Powder | |
| Ni(HCO2)2·2H2O | |
| In the range 180-200° decomposes to Ni,CO,CO2,H2,H2O,CH4. Partially soluble in water | |
| Green powder | |
| 2.15 g/cm3 |
Safety and Handling
Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H341-H350-H360-H372
DOTInformation : Hazard Class: 9, Packing Group: III
EINECSNumber : 222-101-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only