missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) fluoride tetrahydrate, 98+%

Catalog No. p-7049359
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1413218 50 g
AA1413230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1413218 Supplier Thermo Scientific Chemicals Supplier No. 01413218
Only null left

CAS: 13940-83-5

Nickel(II) fluoride tetrahydrate is used in surface treatment of aluminum profile and printing ink making, fluorescence lamp, fluidizer and catalyzer for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13940-83-5
MDL Number MFCD00016263

Specifications

Physical Form Powder
Quantity 50 g
UN Number UN3288
Solubility Information Soluble in water,inorganic acid.
Formula Weight 168.77 (96.71 Anhydrous)
Percent Purity ≥98%
Odor Odorless
Chemical Name or Material Nickel(II) fluoride tetrahydrate
Hazard Category H317-H334-H341-H350-H360-H372
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 233-071-3
RTECSNumber QR6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only