missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) fluoride, anhydrous, 97%

Catalog No. p-7049156
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1306709 10 g
AA1306718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1306709 Supplier Thermo Scientific Chemicals Supplier No. 01306709
Only null left

CAS: 10028-18-9

Nickel fluoride comprises the passivating surface that forms on nickel alloys, e.g. monel, which is why such materials are good to store or transport hydrogen fluoride or elemental fluorine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10028-18-9
MDL Number MFCD00016263

Specifications

Color Yellow
Physical Form Crystalline
Quantity 10 g
UN Number UN3288
Merck Index 14,6508
Solubility Information Slightly soluble in water
Formula Weight 96.71
Percent Purity 97%
Odor Odorless
Sensitivity Hygroscopic
Density 4.72 g/mL
Chemical Name or Material Nickel(II) fluoride, Anhydrous
Show More Show Less
Hazard Category H317-H334-H341-H350i-H360D-H372
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 233-071-3
RTECSNumber QR6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only