missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Dibutyldithiocarbamate 97.0+%, TCI America™

Catalog No. D178125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D178125G 25 g
D1781500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D178125G Supplier TCI America Supplier No. D178125G
Only null left

Chemical Identifiers

CAS 13927-77-0
Molecular Formula C18H36N2NiS4
Molecular Weight (g/mol) 467.433
MDL Number MFCD00067273
InChI Key HPOWMHUJHHIQGP-UHFFFAOYSA-L
Synonym Dibutyldithiocarbamic Acid Nickel(II) Salt, Nickel(II) Bis(dibutyldithiocarbamate)
PubChem CID 6101499
IUPAC Name N,N-dibutylcarbamodithioate;nickel(2+)
SMILES CCCCN(CCCC)C(=S)[S-].CCCCN(CCCC)C(=S)[S-].[Ni+2]

Specifications

Melting Point 89°C
Quantity 25 g
Formula Weight 467.43
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nickel(II) Dibutyldithiocarbamate
EINECSNumber (2)-1247
RTECSNumber QR6140000
TSCA Yes