missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Chloride Hexahydrate, puriss. p.a., ≥98%, Honeywell Fluka™

Catalog No. p-7120617
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045960 1 kg Poly Bottle
60045961 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045960 Supplier Honeywell Chemicals Supplier No. 314621KG
Only null left

Puriss. p.a.,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7791-20-0
MDL Number MFCD00149809

Specifications

Melting Point 80°C
Quantity 1 kg
Linear Formula NiCl2 · 6H2O
UN Number UN3288
Formula Weight 237.69g/mol
Percent Purity ≥98%
Grade puriss. p.a.
Packaging Poly Bottle
Density 3.550g/cm3
Chemical Name or Material Nickel(Ii) Chloride Hexahydrate
Hazard Statement Missing Phrase - N15.00950417-P201-P260-P280-P304 + P340 + P312-P308 + P313
Precautionary Statement H301 + H331-H315-H317-H334-H341-H350i-H360D-H372-H410
DOTInformation Class 6.1 - PG 3 - EHS - RQ - Toxic solid, inorganic, n.o.s., HI: all
RTECSNumber QR6480000