missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) Chloride Hexahydrate, puriss. p.a., ≥98%, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Melting Point | 80°C |
| Quantity | 1 kg |
| Linear Formula | NiCl2 · 6H2O |
| UN Number | UN3288 |
| Formula Weight | 237.69g/mol |
| Percent Purity | ≥98% |
| Grade | puriss. p.a. |
| Packaging | Poly Bottle |
| Density | 3.550g/cm3 |
| Chemical Name or Material | Nickel(Ii) Chloride Hexahydrate |
Safety and Handling
| Hazard Statement | Missing Phrase - N15.00950417-P201-P260-P280-P304 + P340 + P312-P308 + P313 |
| Precautionary Statement | H301 + H331-H315-H317-H334-H341-H350i-H360D-H372-H410 |
| DOTInformation | Class 6.1 - PG 3 - EHS - RQ - Toxic solid, inorganic, n.o.s., HI: all |
| RTECSNumber | QR6480000 |