missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride hexahydrate, Puratronic™, 99.999% (metals basis)

Catalog No. p-4824840
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4318514 25 g
AA4318522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4318514 Supplier Thermo Scientific Chemicals Supplier No. 04318514

CAS: 7791-20-0 | Cl2Ni·6H2O

Used in the manufacture of all kinds of nickel compounds, preservatives and chemical reaction catalyst. Also used in electroplating, analytical chemistry, used for analysis reagent and in gas masks to absorb NH3. It has also been utilized in environmental studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7791-20-0
Molecular Formula Cl2Ni·6H2O
MDL Number MFCD00149809

Specifications

Melting Point 140°C-H2O
Color Green
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.999% (Metals Basis)
Solubility Information Soluble in water (2540g/L) and ethanol.
Formula Weight 237.71 (129.60 Anhydrous)
Odor Odorless
Density 1.92
Chemical Name or Material Nickel(II) chloride hexahydrate
Hazard Category H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 231-743-0
RTECSNumber QR6480000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only