missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) Chloride Anhydrous 98.0+%, TCI America™
Supplier: TCI America N0850500G
Specifications
| Nickel(II) Chloride Anhydrous | |
| Crystalline Powder | |
| Cl2Ni | |
| 3288 | |
| Cl[Ni]Cl | |
| 129.593 | |
| CHEBI:34887 | |
| ≥98.0% (T) |
| 7718-54-9 | |
| 500 g | |
| MFCD00011142 | |
| QMMRZOWCJAIUJA-UHFFFAOYSA-L | |
| dichloronickel | |
| 24385 | |
| 129.59 |
Chemical Identifiers
| 7718-54-9 | |
| 129.593 | |
| QMMRZOWCJAIUJA-UHFFFAOYSA-L | |
| CHEBI:34887 | |
| Cl[Ni]Cl |
| Cl2Ni | |
| MFCD00011142 | |
| 24385 | |
| dichloronickel |
Safety and Handling
EINECSNumber : (1)-0242
RTECSNumber : QR6475000
TSCA : Yes