missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Chloride Anhydrous 98.0+%, TCI America™

Catalog No. N085025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N085025G 25 g
N0850500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N085025G Supplier TCI America Supplier No. N085025G

Chemical Identifiers

CAS 7718-54-9
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.593
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887
IUPAC Name dichloronickel
SMILES Cl[Ni]Cl

Specifications

Physical Form Crystalline Powder
Quantity 25 g
UN Number 3288
Formula Weight 129.59
Percent Purity ≥98.0% (T)
Chemical Name or Material Nickel(II) Chloride Anhydrous
EINECSNumber (1)-0242
RTECSNumber QR6475000
TSCA Yes