missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride, 99.99%, (trace metal basis), anhydrous, powder

CAS: 7718-54-9 | Cl2Ni | 129.6 g/mol

$294.94 - $294.94

Chemical Identifiers

CAS 7718-54-9
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.6
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887
IUPAC Name dichloronickel
SMILES Cl[Ni]Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC378350050
SDS View Documents
Thermo Scientific Chemicals
378350050
5 g Glass bottle
Each for $294.94
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications

Chemical Identifiers

7718-54-9
129.6
QMMRZOWCJAIUJA-UHFFFAOYSA-L
CHEBI:34887
Cl[Ni]Cl
Cl2Ni
MFCD00011142
24385
dichloronickel

Specifications

1001.0°C
Brown to Yellow
5 g
Cl2Ni
MFCD00011142
Solubility in water: slightly soluble
Cl[Ni]Cl
129.6
CHEBI:34887
99.99%
Glass bottle
7718-54-9
Liquid
44.6 to 46% (Ni) (Complexometry)
NiCl2
15, 6590
QMMRZOWCJAIUJA-UHFFFAOYSA-L
dichloronickel
24385
129.6
Trace Metal Basis
Nickel(II) chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.<br

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for

GHS Signal Word: Danger

EINECSNumber : 231-743-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only