missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride, 98%

Catalog No. p-3527073
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC387320500 50 g Glass bottle
AC387322500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC387320500 Supplier Thermo Scientific Chemicals Supplier No. 387320500
Only null left

Nickel(II) chloride, 98%, Cl2Ni, CAS Number-7718-54-9, 250g, 97.5% min. (Argentometry), 100.0, CHEBI:34887, Brown to Orange, 231-743-, 129.6, GHS Signal Word: Danger | CAS: 7718-54-9 | Cl2Ni | 129.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7718-54-9
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887

Specifications

Melting Point 1001.0°C
Color Brown to Orange
CAS Min % 97.5
CAS Max % 100.0
Physical Form Powder
Quantity 50 g
Assay Percent Range 97.5% min. (Argentometry)
Molecular Formula Cl2Ni
Linear Formula NiCl2
Merck Index 15, 6590
Solubility Information Solubility in water: slightly soluble
SMILES Cl[Ni]Cl
IUPAC Name dichloronickel
Molecular Weight (g/mol) 129.6
Formula Weight 129.6
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Nickel(II) chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.<br
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for
EINECSNumber 231-743-

RUO – Research Use Only