missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) carbonate (basic) hydrate, Ni 40% min, typically 99.5% (metals basis)

Catalog No. AA40106A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40106A1 1 kg
AA4010630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40106A1 Supplier Thermo Scientific Chemicals Supplier No. 040106A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nickel Plating pH Adjuster, Catalysts, Ceramic Colors and Glazes | CAS: 155775-31-8 | CH4Ni3O7 | 304.12 g/mol

Nickel plating pH adjuster, catalysts, ceramic colors and glazes.Nickel(II) carbonate (basic) hydrate is used as a precursor to catalysts and in ceramic applications. It acts as an intermediate in the hydrometallurgical purification of nickel from its ores. It is also used in electroplating of nickel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 155775-31-8
Molecular Formula CH4Ni3O7
Molecular Weight (g/mol) 304.12
MDL Number MFCD00016262
InChI Key ATYNVYNPWGEMMI-UHFFFAOYSA-H
Synonym nickel 3+ ;carbonate;hydroxide;hydrate, nickel carbonate, basic hydrate trace metals basis
PubChem CID 86176052
IUPAC Name trinickel(2+) tetrahydroxide carbonate
SMILES [OH-].[OH-].[OH-].[OH-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O

Specifications

Color Green
Physical Form Powder
Quantity 1 kg
Odor Odorless
Appearance Green powder
Assay Percent Range 99.5% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 304.18 (Anhydrous)
Chemical Name or Material Nickel(II) carbonate (basic) hydrate
Hazard Category H302+H332-H315-H317-H319-H334-H341-H350i-H360D-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 240-408-8
RTECSNumber QR6260000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only