missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) carbonate, anhydrous, 98%

Catalog No. p-4782026
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2289718 50 g
AA2289730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2289718 Supplier Thermo Scientific Chemicals Supplier No. 02289718
Only null left

CAS: 3333-67-3 | CNiO3 | 118.70 g/mol

Nickel(II) carbonate is used in sulphamate baths, metal phosphating, electroplating and ceramic applications. It acts as a precursor to catalysts and an intermediate in the hydrometallurgical purification of nickel from its ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3333-67-3
Molecular Formula CNiO3
Molecular Weight (g/mol) 118.70
MDL Number MFCD00011144
InChI Key ZULUUIKRFGGGTL-UHFFFAOYSA-L
Synonym nickel carbonate, nickelous carbonate, nickel ii carbonate, nickel monocarbonate, unii-wm50squ829, carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate nico3
PubChem CID 18746
IUPAC Name nickel(2+);carbonate
SMILES [Ni++].[O-]C([O-])=O

Specifications

Color Beige
Physical Form Powder
Quantity 50 g
Assay Percent Range 98%
Solubility Information Soluble in dilute acids. Insoluble in cold water.
Formula Weight 118.72
Appearance Light powder
Chemical Name or Material Nickel(II) carbonate, anhydrous
Hazard Category H302+H332-H315-H317-H319-H334-H341-H350i-H360D-H372
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Hazard Class: 9, Packing Group: III
EINECSNumber 222-068-2
RTECSNumber QR6200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only