missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) bromide trihydrate, 98%

Catalog No. AA4024230
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4024230 250 g
AA4024218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4024230 Supplier Thermo Scientific Chemicals Supplier No. 04024230

CAS: 7789-49-3 | Br2H6NiO3 | 272.546 g/mol

It is used in Soporific and sedatives pharmaceutical Industry, Photosensitive industry, Analytical Chemistry, and also electrolyte in high energy battery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-49-3
Molecular Formula Br2H6NiO3
Molecular Weight (g/mol) 272.546
MDL Number MFCD00011141
InChI Key UQPSGBZICXWIAG-UHFFFAOYSA-L
Synonym nickel ii bromide trihydrate, dibromonickel trihydrate, nickel dibromide hydrate, nickel bromide trihydrate, br2ni.3h2o, ksc497q6j, dibromonickel-water 1/3, nickel ii bromide trihydrate at
PubChem CID 53471882
IUPAC Name dibromonickel;trihydrate
SMILES O.O.O.[Ni](Br)Br

Specifications

Melting Point 300°C (in -3H2O)
Physical Form Crystalline Granules
Quantity 250 g
UN Number UN3077
Merck Index 14,6502
Solubility Information Soluble in water, alcohol
Formula Weight 272.58 (218.53 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Nickel(II) bromide trihydrate
Show More Show Less
Hazard Category H317-H334-H341-H350-H360-H372
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 236-665-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only