missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) acetate tetrahydrate, 99%, extra pure

Catalog No. AC415540050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415540050 5 g Glass Bottle
AC415541000 100 g Plastic bottle
AC415545000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC415540050 Supplier Thermo Scientific Chemicals Supplier No. 415540050

CAS: 6018-89-9 | C4H14NiO8 | 248.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6018-89-9 , 373-02-4
Molecular Formula C4H14NiO8
Molecular Weight (g/mol) 248.84
MDL Number MFCD00066973
InChI Key OINIXPNQKAZCRL-UHFFFAOYSA-L
Synonym nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
PubChem CID 62601
IUPAC Name nickel(2+);diacetate;tetrahydrate
SMILES O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 250.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Packaging Glass Bottle
Linear Formula (CH3CO2)2Ni·4H2O
Quantity 5 g
Fieser 14,288; 16,308
Merck Index 14, 6498
Solubility Information Solubility in water: 182g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 248.86
Percent Purity 99%
Grade Extra Pure
Physical Form Powder
Chemical Name or Material Nickel(II) acetate tetrahydrate, Extra Pure
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g

RUO – Research Use Only