missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) acetate tetrahydrate, 99+%, for analysis
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 250.0°C |
| Color | White |
| Physical Form | Crystalline Powder, Crystals or Flakes |
| Quantity | 500 g |
| Infrared Spectrum | Authentic |
| Packaging | Plastic bottle |
| Assay Percent Range | 99% min. (Complexometry) |
| Linear Formula | (CH3CO2)2Ni·4H2O |
| Fieser | 14,288; 16,308 |
| Merck Index | 14,6498 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. May cause an allergic skin reaction. Harmful if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Suspected of causing genetic defects. May cause |
| Precautionary Statement | GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective g |
RUO – Research Use Only