Learn More
Nickel(II) acetate tetrahydrate, 98+%
CAS: 6018-89-9 | C4H14NiO8 | 248.84 g/mol
$69.37 - $511.95
Chemical Identifiers
| CAS | 6018-89-9 |
|---|---|
| Molecular Formula | C4H14NiO8 |
| Molecular Weight (g/mol) | 248.84 |
| MDL Number | MFCD00066973 |
| InChI Key | OINIXPNQKAZCRL-UHFFFAOYSA-L |
| Synonym | nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate |
| PubChem CID | 62601 |
| IUPAC Name | nickel(2+);diacetate;tetrahydrate |
| SMILES | O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1302622
|
Thermo Scientific Chemicals
A1302622 |
100 g |
Each for $69.37
|
|
|||||
|
AAA1302636
|
Thermo Scientific Chemicals
A1302636 |
500 g |
Each for $150.41
|
|
|||||
|
AAA130260E
|
Thermo Scientific Chemicals
A130260E |
2500 g |
Each for $511.95
|
|
|||||
Description
Nickel(II) acetate tetrahydrate is used in electroplating. It is also used in the preparation of polynuclear-nickel polyoxotungstate cluster compounds, which are used in designing molecular magnets. Further, it is used as a catalyst and mordant for dyeing textiles, as a sealant for anodized aluminum and to make other nickel compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNickel(II) acetate tetrahydrate is used in electroplating. It is also used in the preparation of polynuclear-nickel polyoxotungstate cluster compounds, which are used in designing molecular magnets. Further, it is used as a catalyst and mordant for dyeing textiles, as a sealant for anodized aluminum and to make other nickel compounds.
Solubility
Soluble in water and methanol. Insoluble in diethyl ether and n-octanol.
Notes
Hygroscopic. Incompatible with oxidizing agents, strong acids and strong bases.
Chemical Identifiers
| 6018-89-9 | |
| 248.84 | |
| OINIXPNQKAZCRL-UHFFFAOYSA-L | |
| 62601 | |
| O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O |
| C4H14NiO8 | |
| MFCD00066973 | |
| nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate | |
| nickel(2+);diacetate;tetrahydrate |
Specifications
| 6018-89-9 | |
| 1.744 | |
| C4H14NiO8 | |
| 100 g | |
| 3730764 | |
| nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate | |
| OINIXPNQKAZCRL-UHFFFAOYSA-L | |
| nickel(2+);diacetate;tetrahydrate | |
| 62601 | |
| ≥98% |
| (decomposition) | |
| Light | |
| MFCD00066973 | |
| UN3077 | |
| 14,6499 | |
| Soluble in water and methanol. Insoluble in diethyl ether and n-octanol. | |
| O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O | |
| 248.84 | |
| 248.84 (176.79 Anhydrous) | |
| Nickel(II) acetate tetrahydrate |
Safety and Handling
GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
H302+H332-H317-H334-H335-H341-H350i-H360D-H372
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 206-761-7
RTECSNumber : QR6126000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only