missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) acetate tetrahydrate, 98+%

Catalog No. AAA1302636
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1302636 500 g
AAA1302622 100 g
AAA130260E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1302636 Supplier Thermo Scientific Chemicals Supplier No. A1302636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6018-89-9 | C4H14NiO8 | 248.84 g/mol

Nickel(II) acetate tetrahydrate is used in electroplating. It is also used in the preparation of polynuclear-nickel polyoxotungstate cluster compounds, which are used in designing molecular magnets. Further, it is used as a catalyst and mordant for dyeing textiles, as a sealant for anodized aluminum and to make other nickel compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nickel(II) acetate tetrahydrate is used in electroplating. It is also used in the preparation of polynuclear-nickel polyoxotungstate cluster compounds, which are used in designing molecular magnets. Further, it is used as a catalyst and mordant for dyeing textiles, as a sealant for anodized aluminum and to make other nickel compounds.

Solubility
Soluble in water and methanol. Insoluble in diethyl ether and n-octanol.

Notes
Hygroscopic. Incompatible with oxidizing agents, strong acids and strong bases.

Chemical Identifiers

CAS 6018-89-9
Molecular Formula C4H14NiO8
Molecular Weight (g/mol) 248.84
MDL Number MFCD00066973
InChI Key OINIXPNQKAZCRL-UHFFFAOYSA-L
Synonym nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
PubChem CID 62601
IUPAC Name nickel(2+);diacetate;tetrahydrate
SMILES O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point (decomposition)
Density 1.744
Odor Light
Quantity 500 g
UN Number UN3077
Beilstein 3730764
Merck Index 14,6499
Solubility Information Soluble in water and methanol. Insoluble in diethyl ether and n-octanol.
Formula Weight 248.84 (176.79 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Nickel(II) acetate tetrahydrate
Show More Show Less
Hazard Category H302+H332-H317-H334-H335-H341-H350i-H360D-H372
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 206-761-7
RTECSNumber QR6126000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only