missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Diethyldithiocarbamate 97.0+%, TCI America™
$53.90 - $363.85
Chemical Identifiers
| CAS | 14267-17-5 |
|---|---|
| Molecular Formula | C10H20N2NiS4 |
| Molecular Weight (g/mol) | 355.217 |
| MDL Number | MFCD00137706 |
| InChI Key | NCLUCMXMAPDFGT-UHFFFAOYSA-L |
| Synonym | Diethyldithiocarbamic Acid Nickel Salt |
| PubChem CID | 84307 |
| IUPAC Name | N,N-diethylcarbamodithioate;nickel(2+) |
| SMILES | CCN(CC)C(=S)[S-].CCN(CC)C(=S)[S-].[Ni+2] |
Chemical Identifiers
| 14267-17-5 | |
| 355.217 | |
| NCLUCMXMAPDFGT-UHFFFAOYSA-L | |
| 84307 | |
| CCN(CC)C(=S)[S-].CCN(CC)C(=S)[S-].[Ni+2] |
| C10H20N2NiS4 | |
| MFCD00137706 | |
| Diethyldithiocarbamic Acid Nickel Salt | |
| N,N-diethylcarbamodithioate;nickel(2+) |
Specifications
| 14267-17-5 | |
| MFCD00137706 | |
| Diethyldithiocarbamic Acid Nickel Salt | |
| CCN(CC)C(=S)[S-].CCN(CC)C(=S)[S-].[Ni+2] | |
| 355.217 | |
| 355.22 | |
| Crystalline Powder |
| C10H20N2NiS4 | |
| 25 g | |
| NCLUCMXMAPDFGT-UHFFFAOYSA-L | |
| N,N-diethylcarbamodithioate;nickel(2+) | |
| 84307 | |
| ≥97.0% (T) | |
| Nickel Diethyldithiocarbamate |
Safety and Handling
EINECSNumber : (2)-1842
TSCA : Yes