missing translation for 'onlineSavingsMsg'
Learn More

Nickel Diethyldithiocarbamate 97.0+%, TCI America™

Catalog No. D2879500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2879500G 500 g
D287925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2879500G Supplier TCI America Supplier No. D2879500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14267-17-5
Molecular Formula C10H20N2NiS4
Molecular Weight (g/mol) 355.217
MDL Number MFCD00137706
InChI Key NCLUCMXMAPDFGT-UHFFFAOYSA-L
Synonym Diethyldithiocarbamic Acid Nickel Salt
PubChem CID 84307
IUPAC Name N,N-diethylcarbamodithioate;nickel(2+)
SMILES CCN(CC)C(=S)[S-].CCN(CC)C(=S)[S-].[Ni+2]

Specifications

Quantity 500 g
Formula Weight 355.22
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nickel Diethyldithiocarbamate
EINECSNumber (2)-1842
TSCA Yes