missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Diethyldithiocarbamate 97.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Formula Weight | 355.22 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Nickel Diethyldithiocarbamate |
Safety and Handling
| EINECSNumber | (2)-1842 |
| TSCA | Yes |