Learn More
United Scientific™ Nickel-Chromium Wire, 4-Ounce Spool
Useful in a wide variety of electricity demonstrations.
Supplier:  United Scientific™ WNC024
 View more versions of this product
Catalog No. S16021
Only null left
Description
Description
  • Bare nickel-chromium connecting wire is supplied in a 4-ounce spool.
  • Sizes Offered in 18, 20, 22, 24 and 28-gauge (SWG)
Specifications
Specifications
Wire
Nickel Chromium
24
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Warning: Cancer and reproductive harm