Learn More
Nickel Chromium wire, 0.25mm (0.01in) dia
CAS: 11106-97-1 | CrNi | 110.69 g/mol
$151.45 - $151.45
Chemical Identifiers
| CAS | 11106-97-1 |
|---|---|
| Molecular Formula | CrNi |
| Molecular Weight (g/mol) | 110.69 |
| MDL Number | MFCD00673110 |
| InChI Key | VNNRSPGTAMTISX-UHFFFAOYSA-N |
| Synonym | nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia |
| PubChem CID | 11320920 |
| IUPAC Name | chromium;nickel |
| SMILES | [Cr].[Ni] |
Description
Nichrome alloys are typically used in resistance wire. They are also used in some dental restorations (fillings) and in other applications. Nichrome is used in a very wide variety of devices where electric heating is required.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 11106-97-1 | |
| 110.69 | |
| VNNRSPGTAMTISX-UHFFFAOYSA-N | |
| 11320920 | |
| [Cr].[Ni] |
| CrNi | |
| MFCD00673110 | |
| nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia | |
| chromium;nickel |
Specifications
| 11106-97-1 | |
| 100 m | |
| Wire | |
| MFCD00673110 | |
| Insoluble in water. | |
| [Cr].[Ni] | |
| 110.69 | |
| Nickel Chromium |
| Silver-colored wire | |
| Odorless | |
| CrNi | |
| nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia | |
| VNNRSPGTAMTISX-UHFFFAOYSA-N | |
| chromium;nickel | |
| 11320920 |
Safety and Handling
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only