missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Chromium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.95% (metals basis), Thermo Scientific Chemicals
$268.33 - $268.33
Chemical Identifiers
| CAS | 11106-97-1 |
|---|---|
| Molecular Formula | CrNi |
| Molecular Weight (g/mol) | 110.69 |
| MDL Number | MFCD00673110 |
| InChI Key | VNNRSPGTAMTISX-UHFFFAOYSA-N |
| Synonym | nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia |
| PubChem CID | 11320920 |
| SMILES | [Cr].[Ni] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 11106-97-1 | |
| 110.69 | |
| VNNRSPGTAMTISX-UHFFFAOYSA-N | |
| 11320920 |
| CrNi | |
| MFCD00673110 | |
| nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia | |
| [Cr].[Ni] |
Specifications
| 11106-97-1 | |
| 50 g | |
| ≈1.7g/slug | |
| CrNi | |
| nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia | |
| VNNRSPGTAMTISX-UHFFFAOYSA-N | |
| chromium nickel | |
| 11320920 | |
| Nickel Chromium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length |
| Ni:Cr; 80:20 wt% | |
| Odorless | |
| Slug | |
| MFCD00673110 | |
| Insoluble in water. | |
| [Cr].[Ni] | |
| 110.69 | |
| 99.95% | |
| (metals basis) |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only