missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chromium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog No. AA4236618
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4236618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4236618 Supplier Thermo Scientific Chemicals Supplier No. 04236618

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 11106-97-1 | Molecular Formula: CrNi | Molecular Weight: 110.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11106-97-1
Molecular Formula CrNi
Molecular Weight (g/mol) 110.69
MDL Number MFCD00673110
InChI Key VNNRSPGTAMTISX-UHFFFAOYSA-N
Synonym nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
PubChem CID 11320920
SMILES [Cr].[Ni]

Specifications

Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr; 80:20 wt%
Quantity 50 g
Odor Odorless
Weight ≈1.7g/slug
Form Slug
Solubility Information Insoluble in water.
IUPAC Name chromium nickel
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Nickel Chromium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
Hazard Category H317-H351
Hazard Statement GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only