missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Chromium slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.5 in.) length, 99.95% (metals basis), Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Avg. Mol. Wt. or Mol. Wt. Range | Ni:Cr; 80:20 wt% |
| Quantity | 10 g |
| Odor | Odorless |
| Weight | ≈3.4g/slug |
| Form | Slug |
| IUPAC Name | chromium;nickel |
| Percent Purity | 99.95% |
| Assay | (metals basis) |
| Chemical Name or Material | Nickel Chromium slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.5 in.) length |
Safety and Handling
| Hazard Category | H317-H351 |
| Hazard Statement | GHS H Statement H351-H317 Suspected of causing cancer. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |