missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chromium lump, 99.9% (metals basis)

Catalog No. AA3629814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3629814 25 g
AA3629822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3629814 Supplier Thermo Scientific Chemicals Supplier No. 03629814

For vacuum deposition | CAS: 11106-97-1 | CrNi | 110.69 g/mol

For vacuum deposition. hey are also used in some dental restorations. Used as Heating Elements, Thermocouples, Wear Resistant Alloys. Used as intermediate and research. Nichrome wire is commonly used in ceramics and Pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11106-97-1
Molecular Formula CrNi
Molecular Weight (g/mol) 110.69
MDL Number MFCD00673110
InChI Key VNNRSPGTAMTISX-UHFFFAOYSA-N
Synonym nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
PubChem CID 11320920
IUPAC Name chromium nickel
SMILES [Cr].[Ni]

Specifications

Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr; 80:20 wt%
Quantity 25 g
Odor Odorless
Particle Size 3 to 12 mm
Form Sintered Lump
Solubility Information Insoluble in water.
Percent Purity 99.9%
Assay Metals basis
Chemical Name or Material Nickel Chromium lump
Hazard Category H317-H351
Hazard Statement GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only