missing translation for 'onlineSavingsMsg'
Learn More

Nickel chromite

CAS: 12018-18-7

$178.30 - $605.49

Chemical Identifiers

CAS 12018-18-7
MDL Number MFCD00064806
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1942414
View Documents
Thermo Scientific Chemicals
01942414
25 g
Each for $178.30
Only null left
 
AA1942422
View Documents
Thermo Scientific Chemicals
01942422
100 g
Each for $605.49
Only null left
 
Description

Description

In mixing Nickel (II) Chromate with Aluminates or Cobalt Chromate, a binary compound known as a spinel is formed that is able to withstand temperatures up to 1000 °C. The products from these mixings can be used as oxidation-resistive components in airplane and space shuttle coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12018-18-7
MFCD00064806

Specifications

12018-18-7
25 g
MFCD00064806
226.7
Nickel chromite
Powder
NiCr2O4
Insoluble in water. Soluble in hydrochloric acid.
Odorless
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350

EINECSNumber : 234-636-7

RTECSNumber : GB9000000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only