missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chloride Hexahydrate, (Crystalline),MP Biomedicals

Cell Culture Reagent

$209.46 - $209.46

Chemical Identifiers

CAS 7791-20-0
Molecular Formula NiCl2 · 6H2O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15582590
View Documents
MP Biomedicals Inc
0215582590
500 g
Each for $209.46
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7791-20-0
NiCl2 · 6H2O

Specifications

7791-20-0
500 g
237.7
Crystalline
NiCl2 · 6H2O
Nickel Chloride Hexahydrate
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'