missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chloride, Hexahydrate, Crystal, 95%, Spectrum™ Chemical

Catalog No. 18611951
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611951 25 g Amber Glass Bottle
18611952 125 g Amber Glass Bottle
18611953 500 g Amber Glass Bottle
18611954 2.5 kg Amber Glass Bottle
18611955 12 kg Poly Pail
18611956 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611951 Supplier Spectrum Chemical Mfg Cor Supplier No. N105025GM

Chemical Identifiers

CAS 7791-20-0
Molecular Formula Cl2H12NiO6
Molecular Weight (g/mol) 237.68
InChI Key LAIZPRYFQUWUBN-UHFFFAOYSA-L
IUPAC Name nickel(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ni++]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 95%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.