missing translation for 'onlineSavingsMsg'
Learn More

Nickel bis(trifluoromethylsulfonyl)imide

CAS: 207861-63-0 | C4F12N2NiO8S4 | 618.964 g/mol

$92.32 - $942.67

Chemical Identifiers

CAS 207861-63-0
Molecular Formula C4F12N2NiO8S4
Molecular Weight (g/mol) 618.964
MDL Number MFCD23380174
InChI Key VFUSIQAHVPEYFD-UHFFFAOYSA-N
Synonym nickel bis-trifluoromethanesulfonimide
PubChem CID 131875825
IUPAC Name bis(trifluoromethylsulfonyl)azanide;nickel(2+)
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ni+2]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA47102MD
View Documents
Thermo Scientific Chemicals
047102MD
250 mg
Each for $92.32
Only null left
 
AA4710203
View Documents
Thermo Scientific Chemicals
04710203
1 g
Each for $241.85
Only null left
 
AA4710206
View Documents
Thermo Scientific Chemicals
04710206
5 g
Each for $942.67
Only null left
 
Description

Description

Nickel(II) Bis(trifluoromethanesulfonyl)imide is used as Surface treatment, Synthesis and catalysis. It is also used as a ionic liquid in synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

207861-63-0
618.964
VFUSIQAHVPEYFD-UHFFFAOYSA-N
131875825
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ni+2]
C4F12N2NiO8S4
MFCD23380174
nickel bis-trifluoromethanesulfonimide
bis(trifluoromethylsulfonyl)azanide;nickel(2+)

Specifications

Green
Solid
C4F12N2NiO8S4
MFCD23380174
nickel bis-trifluoromethanesulfonimide
VFUSIQAHVPEYFD-UHFFFAOYSA-N
bis(trifluoromethylsulfonyl)azanide;nickel(2+)
131875825
Hygroscopic
207861-63-0
250 mg
[(CF3SO2)2N]2Ni
UN3261
It is soluble in water.
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ni+2]
618.964
618.96
Nickel bis(trifluoromethylsulfonyl)imide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H314-H318-H317
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P271-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c

H314-H317-H335-H350

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only